Order Entry
Germany
ContactUsLinkComponent
Substance P ELISA kit
Substance P ELISA kit
Catalog # ENZOADI900018A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Substance P ELISA kit
Catalog # ENZOADI900018A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Alkaline Phosphatase (AP)
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    Substance P
  • Description:
    Substance P ELISA kit
  • Seq:
    02
  • Detection method:
    Colorimetric 405 nm
  • Detection range:
    9,76 - 10000 pg/ml
  • Sensitivity:
    5,3 pg/ml

Specifications

About this item

Highly sensitive Substance P ELISA kit enabling detection of Substance P in plasma, serum, saliva, urine, and supernatants in just 3 hours.

  • Highly sensitive assay, measuring as little as 5.3 pg/ml of Substance P
  • Widely cited in numerous peer reviewed publications
  • High throughput fomat, assay up to 38 samples in duplicate per 96 well kit
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

The Substance P EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit utilized low sample volume and provides the ability to assay up to 38 samples in duplicate per 96 well kit.

Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Caution: RUO - Research Use Only.