Order Entry
Germany
Orders LinkContactUsLinkComponent
 

PGF2α ELISA kit

PGF2α ELISA kit, 1 KIT
Catalog # ENZOADI901069
Supplier:   Enzo Life Sciences
PGF2α ELISA kit
Catalog # ENZOADI901069
Supplier:  Enzo Life Sciences
CAS Number:  
PGF2α ELISA kit

PGF2α ELISA kit, 1 KIT

CAS Number:  
Price
2 060,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

For the quantitative determination of PGF2α in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate and whole blood.

The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors.

 : Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-120 (1x96 wells).

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    PGF2α ELISA kit
  • Cat. No.:
    ENZOADI901069
  • Sensitivity:
    6,71 pg/ml (range 3,05 - 50,000 pg/ml)
  • Target protein:
    PGF2α
  • Assay Type:
    Competitive

Specifications