Order Entry
Germany
ContactUsLinkComponent
24(S)-Hydroxycholesterol ELISA kit
24(S)-Hydroxycholesterol ELISA kit
Catalog # ENZOADI9002100001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
24(S)-Hydroxycholesterol ELISA kit
Catalog # ENZOADI9002100001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    24(S)-Hydroxycholesterol
  • Description:
    24(S)-Hydroxycholesterol ELISA kit
  • Seq:
    02
  • Sensitivity:
    0,78 ng/ml (range 0,39 - 100 ng/ml)

Specifications

About this item

Only quantitative 24(S)-hydroxycholesterol ELISA kit available for neurodegenerative research.

For the quantitative determination of species independent 24(S)-Hydroxycholesterol in serum, plasma, cerebral spinal fluid, culture supernatants and tissue samples.

The 24(S)-Hydroxycholesterol ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 24(S)-Hydroxycholesterol in serum, plasma, tissue culture media (culture supernatant), cerebral spinal fluid and tissue homogenate samples with results in just 2 hours.

Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.