sHLA-G ELISA kit
ENZOALX850309KI01
:
- Pk:1 KIT
- Description:sHLA-G ELISA kit
- Seq:02
- Storage conditions:4 °C (long term)
- Sensitivity:0,6 U/ml (Range 3,91 - 125 U/ml)
For the quantitative determination of soluble human HLA-G (leukocyte antigen-G) in plasma, serum, cell culture supernatant and other body fluids. Cross-reacts strongly with bovine, dog, goat, monkey, rabbit and sheep HLA-G. Cross-reacts weakly with cat HLA-G.