Big endothelin-1 (human), ELISA kit
ENZOADI900022
:
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Endothelin-1
- Description:Big endothelin-1 (Human), ELISA kit, 96 wells
- Seq:02
- Storage conditions:4 °C (long term)
- Detection method:Colorimetric
- Sensitivity:0,23 pg/ml (range 0,78 - 100 pg/ml)
The Big Endothelin-1 (Human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight +1 hour.
- Safe and non-radioactive determination
- Easy to use
- Less steps compared to Western blot
- Reliable and consistent results
For the quantitative determination of human Big ET-1 in culture supernatants and serum. Cited species and sample type include rabbit and lung lavage fluid.
: Kit contains microtiter plate, labeled antibody concentrate, antibody diluent, assay buffer, wash buffer concentrate, standard, TMB substrate, stop solution.