Hydrogen peroxide chemiluminescent detection Kit
Catalog # ENZOADI907012
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:Hydrogen peroxide chemiluminescent detection kit
- Seq:02
- Sensitivity:2,39 ng/ml (range 2,44 - 2,500 ng/ml)
Specifications
About this item
For the quantitative determination of hydrogen peroxide in culture supernatants and cell lysates from any species.
- Immediate results in less than 10 minutes
- Extremely sensitive quantitation of hydrogen peroxide
The Hydrogen Peroxide chemiluminescent detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide. This kit offers more sensitivity over the Hydrogen Peroxide colorimetric detection kit but is not recommended for samples that contain phenol red or hemoglobin.
Ordering information: Contains White Microtiter plate, Standard, Substrate, Trigger solution.