Order Entry
Germany
ContactUsLinkComponent
ROS-ID™ ROS/RNS Detection kit
ROS-ID™ ROS/RNS Detection kit
Catalog # ENZOENZ51001200
Supplier:  Enzo Life Sciences
CAS Number:  
ROS-ID™ ROS/RNS Detection kit
Catalog # ENZOENZ51001200
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    ROS-ID® ROS/RNS detection kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Fluorescence Microscopy
  • Size:
    1 kit

Specifications

About this item

This kit is designed to directly monitor real time reactive oxygen and/or nitrogen species (ROS/RNS) production in live cells using fluorescence microscopy.

Through the combination of the three detection probes with a set of specific inhibitors and activators, this kit enables discrimination among superoxide, nitric oxide and peroxynitrite.

Delivery: Kit contains Nitric Oxide Detection Reagent (Red), 60 µL, Oxidative Stress Detection Reagent (Green), 300 nmoles, Superoxide Detection Reagent (Orange), 300 nmoles, Nitric oxide Inducer (L-Arginine), 100 µL, ROS Inducer (Pyocyanin), 1 µmole, Nitric oxide Scavenger (c-PTIO), 400 nmoles, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and10X Wash Buffer, 15 mL.