Order Entry
Germany
ContactUsLinkComponent
Caspase-3 cell assay kit
Caspase-3 cell assay kit
Catalog # ENZOBMLAK7030001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Caspase-3 cell assay kit
Catalog # ENZOBMLAK7030001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Caspase-3 cell assay kit
  • Seq:
    02
  • Storage conditions:
    –80 °C (long term)
  • Application:
    Colorimetric detection, fluorescent detection, HTS activity assay
  • Size:
    96 well

Specifications

About this item

This kit is a complete assay system for measuring caspase-3 and caspase-3-like activity (DEVDase) in cellular extracts.

It is performed in a 96-well format with all reagents for preparing cell extracts, measuring caspase activity and calibrating the assay. The caspase-3, is useful as a positive control, for testing cellular extracts for endogenous inhibitors, and more

Delivery: Kit includes Caspase-3 enzyme (human, recombinant), pNA Substrate, pNA calibration standard, AMC Substrate, AMC calibration standard (7-amino-4-methylcoumarin), Inhibitor (Ac-DEvD-CHO), Assay buffer, Cell lysis buffer, Half volume microplate.