Hydrogen peroxide fluorometric detection kit
Catalog # ENZOADI907028
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:500 Tests
- Description:Hydrogen peroxide fluorometric detection kit
- Seq:02
- Storage conditions:4 °C (long term)
- Sensitivity:0 - 10 µM (H₂O₂); 0 - 25 mU/ml (peroxidase)
- Size:500 tests
Specifications
About this item
The Hydrogen Peroxide Fluorometric Detection Kit provides fast results in just 10 minutes for the measurement of either hydrogen peroxide or peroxidase activity in a convenient one-step homogeneous assay.
- Flexible in measure hydrogen peroxide or peroxidase activity
- Higher throughput analysis
- Fast results
For the measurement of hydrogen peroxide in cell lysates and culture supernatants from any species.
Delivery: Reaction buffer concentrate, Detection Reagent, Hydrogen peroxide, Horseradish peroxidase