Order Entry
Germany
ContactUsLinkComponent
Catalase fluorometric detection kit
Catalase fluorometric detection kit
Catalog # ENZOADI907027
Supplier:  Enzo Life Sciences
CAS Number:  
Catalase fluorometric detection kit
Catalog # ENZOADI907027
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 Tests
  • Description:
    Catalase fluorometric detection kit
  • Seq:
    02
  • Storage conditions:
    4 °C (long term)
  • Sensitivity:
    0 - 4 U/ml
  • Size:
    500 tests

Specifications

About this item

The Catalase fluorometric detection kit provides a higher throughput sensitive method for measuring catalase activity in just one hour. This is a homogeneous mix-and-read assay that enables you to monitor multiple time points for a kinetic read. Positive control is provided.

  • Easily adaptable to HTS applications
  • Sensitive fluorescent detection system

For the measurement of catalase in cell lysates from any species.

Delivery: Reaction buffer concentrate, Detection Reagent, Horseradish peroxidase concentrate, Hydrogen peroxide, Catalase enzyme