Order Entry
Germany
ContactUsLinkComponent
Anti-HSPA8 Rabbit Polyclonal Antibody
Anti-HSPA8 Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-HSPA8 Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Catalog No:
  • ENZOADISPA819F
  • ENZOADISPA819D
  • Antigen symbol:
  • HSPA8
  • HSPA8
  • Antigen name:
  • Heat Shock Protein Family A (Hsp70) Member 8
  • Heat Shock Protein Family A (Hsp70) Member 8
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • Form:
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified

Specifications