Anti-Dehydrin Rabbit Polyclonal Antibody
Dehydrins are a family of proteins that become abundant during dessication in seedlings and embryos of cereal crop plants including barley, corn, wheat, and rice. Dehydrin proteins have a highly conserved lysine-rich block (KIKEKLPG) found near the carboxy terminus which appears to be characteristic of, and unique to Dehydrins.
- For Western Blot
Recognizes plant Dehydrin. Detects a band of ~65kDa by Western blot.
Type: Primary
Antigen: DEHYDRIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Plant
- Catalog No:
- ENZOADIPLA100F
- ENZOADIPLA100D
- Antigen name:
- Dehydrin
- Dehydrin
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Form:
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Purification:
- Protein A affinity purified
- Protein A affinity purified