
Sulphuric acid 1 M in aqueous solution for analysis of silicates
Catalog # KRAF01888.6020
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
Specification Test Results
- Molarity:~1 M
Specification Test Results
About this item
:
:98.08 g/mol
UN:2796
ADR:,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589