Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sodium hydroxide solution 33% for analysis

1 L
  KRAF21272.3000
 
Sodium hydroxide solution 33% for analysis
  KRAF21272.3000
 :  1310-73-2
Sodium hydroxide solution 33% for analysis

1 L

 :  1310-73-2
 
62,50 €
   
1 * 1 L
 
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :1824
  •  :8,II
 

 

  • Assay:
    33%
  • Form:
    Solution

 

  • Pk:
    1 L