Order Entry
Germany
ContactUsLinkComponent
Homocysteine detection tube, VACUETTE®
Homocysteine detection tube, VACUETTE®
Catalog # GREI454421_1200
Supplier:  Greiner Bio-One
Homocysteine detection tube, VACUETTE®
Catalog # GREI454421_1200
Supplier:  Greiner Bio-One

Specifications

  • Ø:
    13 mm
  • Volume:
    2 ml
  • Description:
    Sandwich tube, Premium (safety twist cap), Vacuette®
  • Length:
    75 mm
  • Cap colour:
    White
  • Pk:
    1.200

Specifications

About this item

This tube is used for determination of the homocysteine concentration in plasma.