Bottles, wide neck, LDPE
Supplier: SEMADENI
Certificates
About this item
These wide mouth bottles are made of soft polyethylene.
- Good chemical resistance, heat resistant up to +80 °C
- Leakproof closure, so they are suitable for use with liquids
- Flexible, excellent impact resistance
Can be used anywhere in the laboratory and for collecting samples. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.