Order Entry
Germany
ContactUsLinkComponent
 
Vial crusher, national
  NSCAC4040-VC
Vial crusher, national
  NSCAC4040-VC

 

  • Description:
    Benchtop vial crusher, manual, 216×597×622,3 mm
  • Pk:
    1

 

 

Breaks and separates liquid from all 1 ml to 100 ml vials and ampoules.

  • Capable of processing approximately 5000 of 2 ml vials per hour
  • Reduces overall waste disposal costs by reducing the total volume of waste

Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapours. Environmentally responsible waste handling, crushed glass is automatically separated from solvent for separate disposal.