PUC18 plasmid DNA
Catalog # SIALD4154-10UG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 µG
- Description:Plasmid DNA, PUC18
Specifications
About this item
Plasmids are isolated from host strain E. coli RRI. These plasmids confer ampicillin resistance and complement defects in β-galactosidase in appropriate host strains.
- Buffered aqueous solution
- Store at −20 °C
Delivery: Plasmid DNA is supplied in a solution of 10 mM Tris-HCl, pH 8,0, with 1 mM EDTA.