


Ethylene oxide (oxirane) solution 2.5-3.3 M in THF
Supplier: Merck
Certificates
About this item
CAS Number:75-21-8
:
Formula:C₂H₄O
MDL Number:MFCD00014482
:44.05 g/mol
Flash Pt:–17 °C
EINECS:200-849-9
UN:2056
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Molarity (in THF):2,5 – 3,3 M