Order Entry
Germany
ContactUsLinkComponent
Surgical Mask, PP
Surgical Mask, PP
Catalog # MENS29197
Supplier:  MENSCH FRANZ
undefined
Surgical Mask, PP
Catalog # MENS29197
Supplier:  MENSCH FRANZ

Specifications

  • Type:
    With elastic bands
  • Colour:
    Green
  • Size:
    175×95 mm
  • Pk:
    1.000

Specifications

About this item

Face mask with elastic bands made from non woven PP, 3-ply (inner and outer layer) and a meltblown fabric filter media.

  • Excellent filter performance >98% BFE
  • Non sterile, lint free, skin-friendly
  • Integrated nose clip, anatomically adjustable
  • Single use
  • Suitable for medical use

No ear pressure even when worn for long periods.

Certifications: EN 14683 Typ II, food safe.