ContactUsLinkComponent
Spheres for water baths, Clifton®
Spheres for water baths, Clifton®
  NICKBP0368
undefined
Spheres for water baths, Clifton®
  NICKBP0368
  • Ø:
    20 mm
  • Description:
    Spheres for water baths, Clifton®
  • Pk:
    200
  • Weight:
    250 g
Hollow spheres are ideal for use as a blanket on the top of open tanks of water. A single layer of spheres covers 91% of the liquid surface trapping air in the gaps which adds to the thermal insulation properties. Spheres have 20 mm Ø.
  • Reduces thermal losses
  • Reduces steaming and splashing