HT (hypoxanthine, thymidine) media, cellgro™, Corning®
MDTC25-047-CI
: Corning
:
- Media Format:Liquid
- Pk:1
- Description:HT (Hypoxanthine, Thymidine)
- Size:100 ml
- Storage temperature:2...8 °C
- Shelf life:20 months
HT media is a 'rescue media' that provides pre-formed purines and pyrimidines to overcome the effects of residual intracellular aminopterin.
- Complete Certificate of Analysis available for each production lot, along with full formulation information
- Produced under the highest industry standards to ensure superior results