Autoclavable cleanroom goggles, Superblast Autoclave
Catalog # BOLESUPBLCLAVE
Supplier: Bollé Safety
Specifications
- Type:Anti-UV, anti-fog, anti-scratch, chemical-resistant
- Colour:Transparent
- Lenses:Clear PC
- Pk:1
Specifications
About this item
These autoclavable cleanroom goggles offer a panoramic vision and an unequalled comfort, with PC lens and PLATINUM® coating included.
- Ventilated frame and high-comfort
- Overflow chute for liquids
- Adjustable strap and clean
- Fits over prescription glasses
- Compatible with half-mask respirator
PLATINUM® coated PC lens: Coating on both sides ensures high scratch-resistance (1.4 cd/m²), high resistance to the most aggressive chemicals and slows the appearance of fogging.
Weight: 92 g