Order Entry
Germany
ContactUsLinkComponent
 
Buffer, triethylammonium acetate, 1 M solution, pH 7,0, Calbiochem®, Millipore®
  625718-1
 :  
Buffer, triethylammonium acetate, 1 M solution, pH 7,0, Calbiochem®, Millipore®
  625718-1
 :  

 

  • Concentration:
    1 M
  • Pack type:
    Plastic ampoule
  • Pk:
    1 L
  • CAS No.:
    5204-74-0
  • Description:
    Triethylammonium acetate buffer
  • Storage:
    15...30 °C
  • Molecular formula:
    C₈H₁₉NO₂
  • Appearance:
    Clear solution
  • pH value:
    7,0

 

 

This buffer solution is used for the purification of chemically synthesised oligonucleotides by HPLC.