Grease, high vacuum, Dow Corning®
Catalog # 44224.KT
Supplier: Thermo Fisher Scientific
Specifications
- Description:Dow Corning® high-vacuum grease, tube, 150 g
- Pk:1
Specifications
About this item
High-vacuum grease is used as an anti-seize grease in ground glass joints and to lubricate synthetic and silicone rubber gaskets. Also used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
- Recommended for vacuums reaching between 1×10-5 and 1×10-6 mmHg
- Immiscible with water, methanol, ethanol, acetone, acetone, glycol and glycerine
- Incompatible with oxidising agents