Order Entry
Germany
Orders LinkContactUsLinkComponent
 

MPI-5a ≥95%

Supplier:   Cayman Chemical
MPI-5a ≥95%
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents

HDAC6 Inhibitor is a potent and selective inhibitor of HDAC6 (IC50= 6 nM) that poorly blocks other HDAC enzymes.

HDAC6 is a predominantly cytoplasmic enzyme that targets α-tubulin, cortactin, and heat shock protein 90, as well as other substrates. In this way, it impacts development, proliferation, invasion, and tumorigenesis. It is cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM.

Synonyms:  N-Hydroxy-2-(1-methyl-1H-pyrrole-2-carbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxamide, MPI_5a
  • CAS Number:1259296-46-2
  • Formula:C₁₆H₁₇N₃O₃
  • MDL Number:MFCD29770783
  • Molecular Weight:299.32 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 81,00 €
    81,00 €
    1 * 500 µG
       81,00 €

  • 1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 141,00 €
    141,00 €
    1 * 1 mg
       141,00 €

  • 5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 580,00 €
    580,00 €
    1 * 5 mg
       580,00 €

  • 10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 000,00 €
    1 000,00 €
    1 * 10 mg
       1 000,00 €