Sleeve protector, cut resistant, Armcut 14 BT
DALL4150057
- Garment application:Mechanical protection
- Gender:Unisex
- Risk area:Cuts
- Colour:Light yellow
- Length:360 mm
- Fabric:Kevlar®
- Pk:1
This 2-ply Kevlar® thread knitted sleeve provides protection of the forearm against cuts.
- Tip of the sleeve stitched like a mitten
- With thumbhole
- Kevlar® fibre offers good cutting resistance and protection against thermal hazards (contact with heat)
- Doubly thick, the cuff's mechanical and thermal performance is greater
- Length of 36 cm ensures vast protection of the forearm
Typical applications include: Automotive and part manufacturing, building and construction, glass industries, iron and steel industry, paper industries, ship building, wood industries.
Gauge: 10
: EN 388 (132X), EN 407 (X1XXXX); Cat. II