Orders LinkContactUsLinkComponent
Dosing spoons, SteriWare®
Dosing spoons, SteriWare®
Supplier:   Sampling Systems
Dosing spoons, SteriWare®
Supplier:  Sampling Systems
Certificates
Dosing spoons, SteriWare®
Dosing spoons, SteriWare®
2 Options Available Below
Product Details & Documents
These disposable and lightweight PS spoons are ideal for sampling and dosing small amounts of powder and liquid. Ideal for work in quality control, laboratories and production.
  • Made from white shatterproof high impact polystyrene (FDA and EU 10/2011 acceptable grade)
  • Handy design comes with a 2,5 ml and 5 ml spoon on either end
  • Available in both sterile and non sterile versions
  • Cross contamination is eliminated - the device can be used straight from the bag
  • Moulded and packed in a cleanroom to assure cleanliness and hygiene
  • BSE/TSE-free
Packaging: Individually bagged and shipped with full documentation including certificates of conformity and sterilisation certificates.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • SAMP8642A-01
    Non sterile
    Non sterile
    2,5/5,0 ml
    Log in for availability
    Log in for availability
    1 * 130 ST
    1 * 130 ST - 57,30 €
    57,30 €
    1 * 130 ST
       57,30 €
  • Sterile
    2,5/5,0 ml
    Log in for availability
    Log in for availability
    1 * 130 ST
    1 * 130 ST - 79,90 €
    79,90 €
    1 * 130 ST
       79,90 €