Anti-RUNX2/Cbfa1 Polyclonal Antibody
Catalog # BSBTPB9158
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:RUNX2/Cbfa1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CBFA1/RUNX2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. CBFA1/RUNX2 information: Molecular Weight: 56648 MW; Subcellular Localization: Nucleus; Tissue Specificity: Specifically expressed in osteoblasts.
Type: Primary
Antigen: RUNX2/Cbfa1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human