Anti-ANP Polyclonal Antibody
Catalog # BSBTPB9295
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:ANP
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for ANP/NPPA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. ANP/NPPA information: Molecular Weight: 16708 MW; Subcellular Localization: Secreted.
Type: Primary
Antigen: ANP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat