pH electrode
Combined pH/temp electrode, with NTC 30 kOhm, Type 230, 0 - 14, Plastic, Gel, BNC
LOVI721230
: Lovibond
Combined pH/temperature electrode with temperature sensor NTC 30 kOhm and BNC connection.
- Connection:BNC
- Type:Type 230
- Description:Combined pH/temp electrode, with NTC 30 kOhm
- Material:Plastic
- Electrolyte:Gel
- pH:0 - 14