ContactUsLinkComponent
Standard, erbium perchlorate solution, for UV wavelength accuracy check
Standard, erbium perchlorate solution, for UV wavelength accuracy check
  WATEWAT042885
 :  WATERS
 :  
Standard, erbium perchlorate solution, for UV wavelength accuracy check
  WATEWAT042885
 :  WATERS
 :  
  • Pk:
    1
  • Description:
    UV wavelength accuracy solution (erbium perchlorate)
 : Kit contains standards appropriate for assessment of UV Detector. 1×10 ml standard solution contains: 10.0 gm/l erbuim perchlorate +/-5%) (purity >=99.9%) in 100% HPLC Grade Water