Orders LinkContactUsLinkComponent
Deuterium lamp
Deuterium lamp
Deuterium lamp, D2, 35 W
Catalog # KNAUA4071
Supplier:   KNAUER
Deuterium lamp
Catalog # KNAUA4071
Supplier:  KNAUER
Deuterium lamp
Deuterium lamp, D2, 35 W
Price
1 030,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This deuterium lamp is suitable for K-2501, P-311 and P-314 photometers.
Specifications
  • Description:
    Deuterium lamp, D2, 35 W