Ethosuximide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-67-8
:
Formula:C₇H₁₁NO₂
MDL Number:MFCD00072123
:141.17 g/mol
Boiling Pt:134 °C (5 mmHg)
Melting Pt:47…50 °C
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or chunks
- Infrared spectrum:Conforms
- Melting point:47 to 52 °C
- Water:≤0.5 %
- HPLC:≥97.5 %