Order Entry
Germany
ContactUsLinkComponent
Valganciclovir hydrochloride 97+%
Health
Valganciclovir hydrochloride 97+%
Catalog # ACRO461270010
CAS Number:  
undefined
Health
Valganciclovir hydrochloride 97+%
Catalog # ACRO461270010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤8 %
  • HPLC:
    ≥97.0 % ( sum of two diastereomers)

Specification Test Results

About this item

 :
 :390.83 g/mol
 :GHS08
Storage Temperature:Refrigerator
CAS Number:175865-59-5
Formula:C₁₄H₂₂N₆O₅·HCl
MDL Number:MFCD08460118