
5-Hydroxy-4-octanone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:496-77-5
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00021928
:144.21 g/mol
Boiling Pt:80…86 °C (12 mmHg)
Flash Pt:71 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %