Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Chloro-1H-pyrazole 98%

4-Chloro-1H-pyrazole 98%

Irritant
4-Chloro-1H-pyrazole 98%
Certificates
4-Chloro-1H-pyrazole 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:15878-00-9
  • Formula:C₃H₃ClN₂
  • MDL Number:MFCD00159634
  • Molecular Weight:102.5200 g/mol
  • Melting Pt:71…73 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to brown Powder or crystalline powder or solid
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 57,40 €
    57,40 €
    1 * 5 g
       57,40 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 172,00 €
    172,00 €
    1 * 25 g
       172,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.