Sodium-L(+)-glutamate monohydrate ≥99%, white powder cell culture reagent
:6106-04-3
:
:C₅H₈NNaO₄·H₂O
:MFCD00150138
:187.13 g/mol
:232 °C (lit.)
:Ambient
- Identity Test:Passes
- Purity:≥99%
- Optical Rotation (c=1, 1 N HCl):+20° to +36°
- Loss on Drying:<0.5%
- pH (5%, aq soln):6.7-7.2
- Solubility (10% aq soln):Clear and Colorless