Order Entry
Germany
ContactUsLinkComponent
 
 
 
 
 
Millon's reagent for the detection of tyrosine
 
undefined
 
 
 
 
Millon's reagent for the detection of tyrosine
 

 

  • Appearance:
    Clear colorless to light yellow Solution
  • Density:
    1.2 to 1.5 kg/l
  • Sensitivity:
    passes test

 

 

Frequently Bought Together