D-(+)-Glucose, anhydrous, white powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Melting Pt:150…170 °C (lit.)
Density:1.544 g/mL
:180.16 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Optical Rotation (c=10, Water, NH4OH):+52.5 ± 5°
- Moisture:≤0.5%