Order Entry
Germany
ContactUsLinkComponent
Florisil® 30-60 mesh for column chromatography
Florisil® 30-60 mesh for column chromatography
Catalog # ACRO457735000
CAS Number:  
undefined
Florisil® 30-60 mesh for column chromatography
Catalog # ACRO457735000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White Powder (May contain black particles)
  • Silicate (as SiO2):
    83.5 to 84.5 %
  • Sodium sulfate (Na2SO4):
    ≤1.0
  • Magnesium oxide (MgO):
    15 to 16 %

Specification Test Results

About this item

 :
 :100.38 g/mol
Storage Temperature:Ambient
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249