

Methyl-3-cyclopentenecarboxylate 97%
Catalog # ACRO457400050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
:
Boiling Pt:149 °C
Flash Pt:49 °C
ADR:3,III
:126.15 g/mol
UN:1993
:GHS02
:GHS05
Storage Temperature:Ambient
CAS Number:58101-60-3
Formula:C₇H₁₀O₂
MDL Number:MFCD04038661