Salbutamol sulphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51022-70-9
:36519-31-0
Formula:(C₁₃H₂₁NO₃)₂·H₂SO₄
MDL Number:MFCD00055200
Melting Pt:180 °C
:576.702 g/mol
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %