Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Amphotericin B 85%

Irritant
Amphotericin B 85%
Certificates
Amphotericin B 85%

2 Options Available Below

Product Details & Documents
Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol
  • Melting Pt:>170 °C
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to orange Powder
  • UV spectrum:
    Complies
  • Water:
    ≤10 %
  • HPLC:
    ≥80 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 134,00 €
    134,00 €
    1 * 1 g
       134,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 524,00 €
    524,00 €
    1 * 5 g
       524,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.