Streptomycin sulphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3810-74-0
:
Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MDL Number:MFCD00037023
:1457.4 g/mol
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Loss on drying:≤7 %
- Sulfate (SO4):18.0 to 21.5