


Ammonium peroxodisulphate (APS) ≥98%, white crystalline powder for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Melting Pt:120 °C (dec., lit.)
Density:950…1050 kg/m³ (bulk density)
UN:1444
ADR:5.1,III
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- pH (1% aq soln):Report Result