1-Naphtholbenzeine
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:145-50-6
:6948-88-5
Formula:C₂₇H₁₈O₂
MDL Number:MFCD00078492
:374.44 g/mol
Melting Pt:230…235 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark red to red-brown to brown-black to orange-brown Powder
- Infrared spectrum:Conforms
- Visual transition interval:from pH 8.2(yellow to orange) to pH 10(green-bleu)
- Molar extinction coefficient:≥50000 (0.01 g/l methanol) (207-213 nm)