
Methyl pentafluorobenzoate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36629-42-2
:
Formula:C₈H₃F₅O₂
MDL Number:MFCD00012172
:226.1002 g/mol
Boiling Pt:87…88 °C (20 mmHg)
Density:1.53 g/cm³
Flash Pt:78 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together