
2,6-Di-tert-butyl-p-benzoquinone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:719-22-2
:
Formula:C₁₄H₂₀O₂
MDL Number:MFCD00001601
:220.31 g/mol
Boiling Pt:75…80 °C (.7 mmHg)
Melting Pt:65…68 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:64 to 70 °C
- GC:≥97.5 %