Order Entry
Germany
Orders LinkContactUsLinkComponent
Methyl orange solution 0,1%

Methyl orange solution 0,1%

250 mL
Catalog # LOVI419436
Supplier:   Lovibond
Methyl orange solution 0,1%
Catalog # LOVI419436
Supplier:  Lovibond
CAS Number:  547-58-0
Methyl orange solution 0,1%

250 mL

CAS Number:  547-58-0
Price
15,50 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
  • CAS Number:547-58-0
  • Formula:C₁₄H₁₄N₃NaO₃S
  • MDL Number:MFCD00007502
  • Molecular Weight:327.34 g/mol

Documents

Specifications

Specifications

  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.